missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Puriss. p.a., ACS Reagent, ≥99.8% (GC), Honeywell Riedel-de Haën™

Product Code. 15637260
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15637260

Brand: Honeywell-Riedel-de Haen 827021L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.8% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -42°C (literature)
CAS 110-86-1
CAS Max % 100.0000%
Density 0.983 g/mL at 20 °C; 0.978 g/mL at 25 °C(lit.)
Boiling Point 115°C
Quantity 1 L
Flash Point 20°C
Packaging Glass Bottle
Molecular Formula C5H5N
Linear Formula C5H5N
MDL Number MFCD00011732
UN Number UN1282
Beilstein 103233
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES C1=CC=NC=C1
IUPAC Name pyridine
Molecular Weight (g/mol) 79.102
PubChem CID 1049
ChEBI CHEBI:16227
Formula Weight 79.1
Percent Purity >99.8%
Grade ACS Reagent
Chemical Name or Material Pyridine, Puriss. p.a., ACS Reagent, ≥99.8% (GC)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.