missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, puriss., Reag. Ph. Eur., Honeywell™

Product Code. 15694710
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. Quantity unitSize
15694710 1 L 1 L
1 options
This item is not returnable. View return policy

Product Code. 15694710

Brand: Honeywell Riedel-de Haën 349451L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 110-86-1
Density 0.978g/mL at 25°C
Quantity 1 L
Explosion Limit 0.124
Flash Point 17°C (62.6°F)
Ignition Point 899°F
Molecular Formula C5H5N
MDL Number MFCD00011732
Vapor Pressure 10 mmHg (13.2°C), 20 mmHg (25°C)
UN Number UN1282
Beilstein 103233
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES C1=CC=NC=C1
IUPAC Name pyridine
Molecular Weight (g/mol) 79.102
PubChem CID 1049
ChEBI CHEBI:16227
Formula Weight 79.1g/mol
Percent Purity ≥99.5% (GC)
Grade Reagent Ph. Eur./puriss.
Chemical Name or Material Pyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.