missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-Sulfur Trioxide Complex, TRC

Product Code. 30412004
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30412004 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30412004

Brand: Toronto Research Chemicals TRCP99156010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyridine-Sulfur Trioxide Complex
CAS 26412-87-3
Molecular Formula C5H5N.SO3
Quantity 10 g
Synonym Sulfur Trioxide-Pyridine, SO3-Py
InChI Formula InChI=1S/C5H5N.O3S/c1-2-4-6-5-3-1;1-4(2)3/h1-5H;
SMILES O=S(=O)=O.c1ccncc1
IUPAC Name pyridine;sulfur trioxide
Molecular Weight (g/mol) 159.16
Formula Weight 158.999
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.