missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium bromide perbromide, 90+%, technical

Product Code. 10583321 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10583321

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39416-48-3 | C5H6Br3N | 319.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39416-48-3
Molecular Formula C5H6Br3N
Molecular Weight (g/mol) 319.83
MDL Number MFCD00013223
Synonym Pyridine hydrobromide, compound with bromine

Specifications

Melting Point 127°C to 133°C
Color Brown to Orange-Red
Infrared Spectrum Authentic
Assay Percent Range 90% min. (Iodometry)
Packaging Glass bottle
Quantity 500 g
Beilstein 20, V,5, 181
Fieser 01,967; 05,568; 06,499; 09,399; 10,333; 14,268; 15,276
Merck Index 15, 8085
Solubility Information Solubility in water: decomposes. Other solubilities: sparingly soluble in acetic acid
Formula Weight 319.83
Percent Purity 90+%
Grade Technical
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Pyridinium bromide perbromide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Pyridinium bromide perbromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.