missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium chlorochromate, 98%

Product Code. 10184150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10184150 25 g Plastic bottle 25g
10012270 100 g Plastic bottle 100g
10194150 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10184150 Supplier Thermo Scientific Acros Supplier No. 183670250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26299-14-9 | C5H6ClCrNO3 | 215.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26299-14-9
Molecular Formula C5H6ClCrNO3
Molecular Weight (g/mol) 215.56
MDL Number MFCD00013106
Synonym PCC

Specifications

Melting Point 205.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Fieser 06,498; 07,308; 08,425; 09,397; 10,334; 11,450; 12,256; 13,237; 14,269; 15,57
Merck Index 15, 8086
Solubility Information Solubility in water: .. Other solubilities: insoluble in dichloromethane
Formula Weight 215.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium chlorochromate, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyridinium chlorochromate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Oxidising solid Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.