missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium dichromate, 98%

Product Code. 11956311 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11956311 1 kg Plastic bottle 1kg
10164310 50 g Plastic bottle 50g
10348050 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11956311 Fournisseur Thermo Scientific Acros Code fournisseur 200880010
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 20039-37-6 | C10H12Cr2N2O7 | 376.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 20039-37-6
Molecular Formula C10H12Cr2N2O7
Molecular Weight (g/mol) 376.2
InChI Key LMYWWPCAXXPJFF-UHFFFAOYSA-P
Synonym pyridinium dichromate, cornforth reagent, pyridinium dichromate pdc, oxido-oxido dioxo chromio oxy-dioxochromium; pyridin-1-ium, pyridiniumdichromate, pyridinium-dichromate, dipyridinium dichromate, bis pyridium chromate, pyridiniumdichromate pdc, acmc-209f5g
PubChem CID 2724130
IUPAC Name oxido-(oxido(dioxo)chromio)oxy-dioxochromium;pyridin-1-ium
SMILES C1=CC=[NH+]C=C1.C1=CC=[NH+]C=C1.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Spécification

Melting Point 152.0°C to 153.0°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Plastic bottle
Quantity 1 kg
Fieser 09,399; 10,335; 12,419; 14,269; 15,57
Merck Index 15,8087
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmf, dmso and dimethylacetamide -fairly, soluble in acetonenitrile - sparingly soluble in, methylene chloride, chloroform and ether - pract., insol.in hexane,toluene,ethyl ether,ethyl acetate
Formula Weight 376.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium dichromate
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Pyridinium dichromate
Mention d'avertissement
  • Danger
Catégorie de danger
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Mention de danger
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H350i-May cause cancer by inhalation.
  • H272-May intensify fire; oxidiser.
  • H228-Flammable solid.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.