missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium p-toluenesulfonate, 98+%

Product Code. 11427677
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427677 25 g 25g
11437677 100 g 100g
11447677 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427677 Supplier Thermo Scientific Alfa Aesar Supplier No. A15708.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24057-28-1 | C12H13NO3S

Pyridinium p-Toluenesulfate is used in the synthesis of a subdomain of the cytochrome P450 BM3 enzyme, for use in screening systems for drug candidates. Efficient catalyst for preparing tetrahydropyranyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridinium p-Toluenesulfate is used in the synthesis of a subdomain of the cytochrome P450 BM3 enzyme, for use in screening systems for drug candidates. Efficient catalyst for preparing tetrahydropyranyl ethers.

Solubility
Soluble in water (partly), ethanol, acetone, dichloromethane, chloroform, methanol and dimethyl sulfoxide.

Notes
Moisture Sensitive. Store in a tightly closed container. Store in a cool, dry, well-ventilated. Store protected from moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24057-28-1
Molecular Formula C12H13NO3S
MDL Number MFCD00013108
Synonym pyridinium p-toluenesulfonate, ppts, pyridin-1-ium 4-methylbenzenesulfonate, pyridinium 4-toluenesulfonate, pyridinium tosylate, pyridinium 4-methylbenzenesulfonate, pyridinium p-toluene sulfonate, pyridinium p-toluene sulphonate, pyridinium para-toluenesulfonate, 4-toluenesulfonic acid pyridine salt

Specifications

Melting Point 116°C to 120°C
Quantity 25 g
Beilstein 3764305
Sensitivity Moisture sensitive
Solubility Information Soluble in water (partly),ethanol,acetone,dichloromethane,chloroform,methanol and dimethyl sulfoxide.
Formula Weight 251.31
Percent Purity ≥98%
Chemical Name or Material Pyridinium p-toluenesulfonate
compliance-icons
Product Identifier
  • Pyridinium-p-toluenesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Pyridinium-p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.