missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium p-toluenesulfonate, 98%

Product Code. 10482401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10482401 25 g Glass bottle 25g
10722233 5 g Glass bottle 5g
10676974 100 g Glass bottle 100g
11966311 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10482401 Supplier Thermo Scientific Acros Supplier No. 213740250
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 24057-28-1 | C12H13NO3S | 251.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24057-28-1
Molecular Formula C12H13NO3S
Molecular Weight (g/mol) 251.3
MDL Number MFCD00013108
Synonym pyridinium p-toluenesulfonate, ppts, pyridin-1-ium 4-methylbenzenesulfonate, pyridinium 4-toluenesulfonate, pyridinium tosylate, pyridinium 4-methylbenzenesulfonate, pyridinium p-toluene sulfonate, pyridinium p-toluene sulphonate, pyridinium para-toluenesulfonate, 4-toluenesulfonic acid pyridine salt

Specifications

Melting Point 116.0°C to 120.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 20, V,5, 160
Fieser 08,427; 09,400; 12,420; 15,276; 16,287
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ethanol, acetone, diethyl ether
Formula Weight 251.3
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Pyridinium p-toluenesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Pyridinium p-toluenesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.