missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridostatin hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Pyridostatin (RR82) hydrochloride is a G-quadruplex DNA stabilizing agent (Kd=490 nM). Pyridostatin hydrochloride promotes growth arrest in human cancer cells by inducing replication- and transcription-dependent DNA damage. Pyridostatin hydrochloride targets the proto-oncogene Src. Pyridostatin hydrochloride reduced SRC protein levels and SRC-dependent cellular motility in human breast cancer cells.
Specifications
Specifications
| Chemical Name or Material | Pyridostatin hydrochloride |
| CAS | 1781882-65-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H37Cl5N8O5 |
| Quantity | 50 mg |
| Synonym | RR82 hydrochloride |
| Solubility Information | H2O : 50 mg/mL (64.19 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC(OCCN)=CC(C(NC2=NC3=C(C(OCCN)=C2)C=CC=C3)=O)=N1)NC4=CC(OCCN)=C5C=CC=CC5=N4.Cl.Cl.Cl.Cl.Cl |
| Molecular Weight (g/mol) | 778.94 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction