missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal 5'-Phosphate, TRC

Product Code. 30003312
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003312 10 g 10g
30004175 25 g 25g
17964925 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003312 Supplier Toronto Research Chemicals Supplier No. TRCP99171510G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-47-7
Molecular Formula C8 H10 N O6 P
Molecular Weight (g/mol) 247.14
Synonym 4-Pyridinecarboxaldehyde, 3-hydroxy-2-methyl-5-[(phosphonooxy)methyl]-, Pyridoxal phosphate (6CI), Pyridoxal, 5-(dihydrogen phosphate) (8CI), 3-Hydroxy-2-methyl-5-[(phosphonooxy)methyl]-4-pyridinecarboxaldehyde, 2-Methyl-3-hydroxy-4-formyl-5-pyridylmethylphosphoric acid, 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde 5-phosphate, Apolon B6, Biosechs, Codecarboxylase, Coenzyme B6, Hairoxal, Hexermin P, Hi-Pyridoxin, Hiadelon, NSC 82388, PAL-P, PLP, Phosphopyridoxal, Phosphopyridoxal coenzyme, Piodel, Pydoxal, Pyridoxal 5-monophosphoric acid ester, Pyridoxal 5-phosphate, Pyridoxal 5'-phosphate, Pyridoxal P, Pyridoxal monophosphate, Pyridoxaldehyde phosphate, Pyridoxyl phosphate, Pyromijin, Sechvitan, Vitahexin P, Vitamin B6 phosphate, Vitamin B6 phosphate (ester), Vitazechs
IUPAC Name (4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate
SMILES Cc1ncc(COP(=O)(O)O)c(C=O)c1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H10NO6P/c1-5-8(11)7(3-10)6(2-9-5)4-15-16(12,13)14/h2-3,11H,4H2,1H3,(H2,12,13,14)
Formula Weight 247.0246
Percent Purity >95%
Chemical Name or Material Pyridoxal-5-Phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.