missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxal hydrochloride, 99%

Product Code. 11423458
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423458 5 g 5g
11433458 25 g 25g
11443458 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423458 Supplier Thermo Scientific Alfa Aesar Supplier No. A17855.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65-22-5 | C8H10ClNO3 | 203.622 g/mol

Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridoxal hydrochloride is used in labeling of amino acids for their detection. It finds application in neurotransmitters, sphingolipids and aminolevulinic acid. It is also used as a nutritional agent. It is also applied in research studies like biochemical, drugs and in media supplements.

Solubility
Soluble in water and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids, strong reducing agents and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-22-5
Molecular Formula C8H10ClNO3
Molecular Weight (g/mol) 203.622
MDL Number MFCD00012809
InChI Key FCHXJFJNDJXENQ-UHFFFAOYSA-N
Synonym pyridoxal hydrochloride, pyridoxal hcl, vitamin b6 hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde hydrochloride, unii-1416kf0qbc, 2-methyl-3-hydroxy-4-formyl-5-hydroxymethylpyridine hydrochloride, pl hcl, 3-hydroxy-5-hydroxymethyl-2-methylisonicotinaldehyde, hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methyl-4-pyridinecarboxaldehyde hydrochloride, 3-hydroxy-5-hydroxymethyl-2-methylpyridine-4-carbaldehyde hydrochloride
PubChem CID 6171
IUPAC Name 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carbaldehyde;hydrochloride
SMILES CC1=NC=C(C(=C1O)C=O)CO.Cl

Specifications

Melting Point 172°C to 174°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3656994
Sensitivity Hygroscopic
Merck Index 14,7978
Solubility Information Soluble in water and ethanol.
Formula Weight 203.63
Percent Purity 99%
Chemical Name or Material Pyridoxal hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.