missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine Cyclic Ether Impurity Hydrochloride Salt, TRC

Product Code. 30009405
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009405 250 mg 250mg
30008713 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009405 Supplier Toronto Research Chemicals Supplier No. TRCP991740250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1006-21-9
Molecular Formula C8 H9 N O2 . Cl H
Molecular Weight (g/mol) 187.62
Synonym 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol Hydrochloride, Pyridoxine Hydrochloride Imp. A (EP) as Hydrochloride
IUPAC Name 6-methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol;hydrochloride
SMILES Cl.Cc1ncc2COCc2c1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H9NO2.ClH/c1-5-8(10)7-4-11-3-6(7)2-9-5;/h2,10H,3-4H2,1H3;1H
Formula Weight 187.04
Percent Purity >95%
Chemical Name or Material 6-Methyl-1,3-dihydrofuro[3,4-c]pyridin-7-ol Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.