missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pyridoxine hydrochloride, 98+%, extra pure

Product Code. 10605972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10605972 50 g Glass bottle 50g
10195303 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10605972 Supplier Thermo Scientific Chemicals Supplier No. 150770500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-56-0 | C8H11NO3·HCl | 205.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H11NO3·HCl
Molecular Weight (g/mol) 205.64
MDL Number MFCD00012807
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
Synonym pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
PubChem CID 6019
ChEBI CHEBI:30961
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
SMILES CC1=NC=C(C(=C1O)CO)CO.Cl

Specifications

Melting Point 214.0°C
Color White
pH 2.4 to 3.0 (5% aq. soln.)
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 50 g
Beilstein 21, V,5, 492
Merck Index 15, 8095
Solubility Information Solution
Formula Weight 205.64
Percent Purity 98+%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Pyridoxine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.