missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine hydrochloride, 98%, MP Biomedicals™

Product Code. 11446142 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
Product Code. Quantity unitSize
11446142 10 g 10g
11466142 50 g 50g
11476142 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11446142

Brand: MP Biomedicals 0210277710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.638
MDL Number MFCD00012807
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
Synonym pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
PubChem CID 6019
ChEBI CHEBI:30961
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
SMILES CC1=NC=C(C(=C1O)CO)CO.Cl

Specifications

Color White
Density 1.43g/mL
Assay Percent Range >98%
Quantity 10 g
Formula Weight 205.64
Percent Purity 0.98
Physical Form Powder
Chemical Name or Material Pyridoxine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.