missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine Hydrochloride, TRC

Product Code. 17940915
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17940915 50 mg 50mg
30007376 1 g 1g
17911185 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17940915 Supplier Toronto Research Chemicals Supplier No. TRCP99173550MG
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8 H11 N O3 . Cl H
Molecular Weight (g/mol) 205.64
Synonym Pyridoxine Hydrochloride, 3,4-Pyridinedimethanol, 5-hydroxy-6-methyl-, hydrochloride (1:1), 3,4-Pyridinedimethanol, 5-hydroxy-6-methyl-, hydrochloride (9CI), Pyridoxol, hydrochloride (7CI,8CI), 5-Hydroxy-6-methyl-3,4-pyridinedimethanol hydrochloride, Pyridoxol hydrochloride, 2-Methyl-3-hydroxy-4,5-bis(hydroxymethyl)pyridine hydrochloride, 2-Methyl-3-hydroxy-4,5-di(hydroxymethyl)pyridine hydrochloride, 3-Hydroxy-4,5-bis(hydroxymethyl)-2-methylpyridine hydrochloride, 3-Hydroxy-4,5-dimethylol-α-picoline hydrochloride, 4,5-Bis(hydroxymethyl)-2-methylpyridin-3-ol hydrochloride, 4,5-Bis(hydroxymethyl)-3-hydroxy-2-methylpyridine hydrochloride, 4,5-Di(hydroxymethyl)-3-hydroxy-2-methylpyridine hydrochloride, 4,5-Dimethylol-3-hydroxy-2-methylpyridine hydrochloride, 5-Hydroxy-6-methyl-3,4-pyridinedicarbinol hydrochloride, 5-Hydroxy-6-methyl-3,4-pyridinedimethanol hydrochloride, Aderomine hydrochloride, Aderoxin, Aderoxine, Becilan, Beesix, Benadon, Bonasanit, Campoviton 6, Hexabetalin, Hexabione hydrochloride, Hexavibex, Hexermin, Hexobion, Pyridipca, Pyridox, Pyridoxin hydrochloride, Pyridoxine chloride, Pyridoxine hydrochloride, Pyridoxine hydrogen chloride, Pyridoxine monohydrochloride, Pyridoxinium chloride, Vitamin B6 hydrochloride
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
SMILES Cl.Cc1ncc(CO)c(CO)c1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H
Formula Weight 205.0506
Percent Purity >95%
Chemical Name or Material Pyridoxine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.