missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine Hydrochloride, U.S.P. - F.C.C., J.T. Baker™

Product Code. 15568114 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
100g
5g
Product Code. Quantity Packaging unitSize
15568114 5 g Wide Mouth Amber Glass Bottle 5g
15578114 100 g Wide Mouth Amber Glass Bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 15568114

Brand: Avantor J.T.Baker 334301

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.638
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
Synonym pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
PubChem CID 6019
ChEBI CHEBI:30961
IUPAC Name 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
SMILES CC1=NC=C(C(=C1O)CO)CO.Cl

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 5 g
Formula Weight 205.64
Grade FCC/USP
Chemical Name or Material Pyridoxine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.