missing translation for 'onlineSavingsMsg'
Learn More

Pyrimethamine, MedChemExpress

Product Code. 16447426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447426 100 mg 100mg
16411896 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16447426 Supplier Medchem Express Supplier No. HY18062
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pyrimethamine(RP4753) is a medication used for protozoal infections; interferes with tetrahydrofolic acid synthesis from folic acid by inhibiting the enzyme dihydrofolate reductase (DHFR).

Pyrimethamine(RP4753) is a medication used for protozoal infections

Specifications

Chemical Name or Material Pyrimethamine
CAS 58-14-0
Color White
Purity Grade Notes Research
Molecular Formula C12H13ClN4
Quantity 100 mg
Synonym Pirimecidan Pirimetamin RP 4753
Solubility Information DMSO : 20 mg/mL (80.41 mM; Need ultrasonic)
SMILES NC1=NC(N)=C(C2=CC=C(Cl)C=C2)C(CC)=N1
Molecular Weight (g/mol) 248.71
Formula Weight 248.71
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.