missing translation for 'onlineSavingsMsg'
Learn More

pyrimidine-5-boronic acid, 97%, Thermo Scientific™

Product Code. 10245804
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10245804 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10245804 Supplier Thermo Scientific Maybridge Supplier No. MAY00076DA
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 321724-19-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H15BN2O2
MDL Number MFCD05155223
Quantity 1 g
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine-5-boronic acid pinacol ester, pinacol ester pyrimidinyl-5-boronic acid, 5-pyrimidineboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidyl-5-boronic acid pinacolate, pyrimidine-5-boronic acid, pinacol ester, pyrimidine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pyrimidin-5-ylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-pyrimidin-5-yl-1,3,2-dioxaborolane
InChI Key WSRGGSAGSUEJKQ-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CN=CN=C1
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Molecular Weight (g/mol) 206.05
PubChem CID 4192667
Formula Weight 206.05
Percent Purity 97%
Chemical Name or Material Pyrimidine-5-boronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.