missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-boronic acid, 98%, Thermo Scientific™

Product Code. 15439757
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439757 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15439757 Supplier Thermo Scientific Alfa Aesar Supplier No. H52984.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Pyrimidine 5-boronic acid could be used in an analogous one-pot copper-catalyzed DBAD coupling and condensation. The widely utilized Suzuki-Miyaura coupling allows for the coupling of boronic acids with carbon halides and surrogates.

Solubility
Soluble in Ether, MeOH, DMSO, THF.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 109299-78-7
Melting Point >300°C
Molecular Formula C4H5BN2O2
MDL Number MFCD03002366
Quantity 250 mg
Synonym 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid
Solubility Information Soluble in Ether,MeOH,DMSO,THF.
InChI Key HZFPPBMKGYINDF-UHFFFAOYSA-N
SMILES OB(O)C1=CN=CN=C1
IUPAC Name pyrimidin-5-ylboronic acid
Molecular Weight (g/mol) 123.91
PubChem CID 2795193
Formula Weight 123.91
Percent Purity 98%
Chemical Name or Material Pyrimidine-5-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.