missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-carboxaldehyde, 97%

Product Code. 15460748
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460748 25 g 25g
15440748 1 g 1g
15450748 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460748 Supplier Thermo Scientific Alfa Aesar Supplier No. H54448.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10070-92-5 | C5H4N2O | 108.10 g/mol

Pyrimidine-5-carboxaldehyde is used in the preparation of (S)- and (R)-pyrimidyl alkanol by reacting with diisopropylzinc and (n-butyl)ethyl(n-hexyl)(n-propyl)methane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine-5-carboxaldehyde is used in the preparation of (S)- and (R)-pyrimidyl alkanol by reacting with diisopropylzinc and (n-butyl)ethyl(n-hexyl)(n-propyl)methane.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10070-92-5
Molecular Formula C5H4N2O
Molecular Weight (g/mol) 108.10
MDL Number MFCD03426065
InChI Key FREJAOSUHFGDBW-UHFFFAOYSA-N
Synonym pyrimidine-5-carboxaldehyde, 5-formylpyrimidine, pyrimidine-5-carboxyaldehyde, 5-pyrimidinecarboxaldehyde, 5-pyrimidinecarbaldehyde, 5-pyrimidinaldehyde, 5-formyl pyrimidine, pubchem5298, pyrimidine-5-aldehyde, pyrimidine 5-carboxaldehyde
PubChem CID 2761034
IUPAC Name pyrimidine-5-carbaldehyde
SMILES O=CC1=CN=CN=C1

Specifications

Melting Point 39°C to 43°C
Density 1.23
Boiling Point 80°C to 82°C (4 mmHg)
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 108.1
Percent Purity 97%
Chemical Name or Material Pyrimidine-5-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.