missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-carboxylic acid, 95%

Product Code. 15477938
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477938 5 g 5g
15487938 250 mg 250mg
15467938 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15477938 Supplier Thermo Scientific Alfa Aesar Supplier No. H61406.06
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4595-61-3 | C5H4N2O2 | 124.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4595-61-3
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00856162
InChI Key IIVUJUOJERNGQX-UHFFFAOYSA-N
Synonym 5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
PubChem CID 78346
IUPAC Name pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=CN=C1

Specifications

Melting Point 259°C to 261°C
Quantity 5 g
Formula Weight 124.1
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Pyrimidine-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.