missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Pyriproxyfen-d4 (pyridyl-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Pyriproxyfen-D4 |
| CAS | 2446366-95-4 |
| Molecular Formula | C20H15D4NO3 |
| Quantity | 0.01 g |
| Synonym | 2-[1-Methyl-2-(4-phenoxyphenoxy)ethoxy]pyridine-D4 |
| InChI Formula | InChI=1 S/C20H19NO3/c1-16(23-20-9-5-6-14-21-20)15-22-17-10-12-19(13-11-17)24-18-7-3-2-4-8-18/h2-14,16 H,15H2,1H3/i5D,6 D,9 D,14 D |
| SMILES | [2 H]C1=NC(OC(C)COC(C=C2)=CC=C2OC3=CC=CC=C3)=C([2 H])C([2 H])=C1[2 H] |
| IUPAC Name | 2-((1-(4-phenoxyphenoxy)propan-2-yl)oxy)pyridine-3,4,5,6-d4 |
| Molecular Weight (g/mol) | 325.39 |
| Formula Weight | 325.16 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction