missing translation for 'onlineSavingsMsg'
Learn More

Pyrithioxin dihydrochloride, MedChemExpress

Product Code. 16438506
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438506 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438506 Supplier Medchem Express Supplier No. HYB0910A
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pyrithioxin dihydrochloride is a neurodynamic compound, combined with a short period of hyperventilation (HV) was applied in cerebral infarct patients with Hemiplegia.

Pyrithioxin dihydrochloride is a neurodynamic compound, combined with a short period of hyperventilation (HV) was applied in cerebral infarct patients with Hemiplegia.

Specifications

Chemical Name or Material Pyrithioxin dihydrochloride
CAS 10049-83-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H22Cl2N2O4S2
Quantity 100 mg
Synonym Pyritinol dihydrochloride Pyridoxine disulfide dihydrochloride Vitamin B6 disulfide dihydrochloride
Solubility Information H2O : ≥ 41 mg/mL (92.89 mM)
SMILES OCC1=C(CSSCC2=C(CO)C(O)=C(C)N=C2)C=NC(C)=C1O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 441.39
Formula Weight 441.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.