missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, ACS, 99%

Product Code. 11379658
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379658 250 g 250g
11369658 10 g 10g
11359658 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379658 Supplier Thermo Scientific Alfa Aesar Supplier No. 044152.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 87-66-1 | C6H6O3 | 126.11 g/mol

Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal- complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

Melting Point 131°C to 135°C
Color White
Density 1.45 g/mL
Boiling Point 309°C
Odor Odorless
Quantity 250 g
UN Number UN2811
Beilstein 907431
Sensitivity Air and light sensitive
Merck Index 14,8000
Solubility Information Soluble in ethanol,water and ether. Slightly soluble in chloroform.
Formula Weight 126.11
Percent Purity 99%
Grade ACS
Physical Form Powder
Chemical Name or Material Pyrogallol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.