missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol Solution, ∽25% in H2O; Honeywell™ Fluka™

Product Code. 15667260 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 15667260

Brand: Honeywell-Fluka 83135250ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 87-66-1
Color Yellow
Density 1.082g/mL at 20°C
Packaging Glass bottle
Molecular Formula C6H6O3
MDL Number MFCD00002192
Quantity 250 mL
UN Number NONH for all modes of transport
Beilstein 907431
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILES OC1=CC=CC(O)=C1O
IUPAC Name benzene-1,2,3-triol
Molecular Weight (g/mol) 126.11
PubChem CID 1057
ChEBI CHEBI:16164
Formula Weight 126.11g/mol
Physical Form Liquid
Chemical Name or Material Pyrogallol solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.