missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, TRC

Product Code. 30137391
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137391 5 g 5g
17904845 25 g 25g
17914655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137391 Supplier Toronto Research Chemicals Supplier No. TRCP9974305G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6 H6 O3
Molecular Weight (g/mol) 126.11
Synonym Pyrogallol (8CI), 1,2,3-Hydroxybenzene, 1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, 2,6-Dihydroxyphenol, Antioxidant PY, Benzene-1,2,3-triol, C.I. 76515, C.I. Oxidation Base 32, Fouramine Brown AP, Fourrine 85, Fourrine PG, Gallol, NSC 5035, Pyrogallic acid, Pyrogallol acid
IUPAC Name benzene-1,2,3-triol
SMILES Oc1cccc(O)c1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H
Formula Weight 126.0317
Percent Purity >95%
Chemical Name or Material Pyrogallol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.