missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Pyrogen-free Test Tubes without Screw Caps

Product Code. 11620311
Change view
Click to view available options
Length (Metric):
100 mm
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11620311 100 mm Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11620311 Supplier Lonza™ Supplier No. N207
  • ONLINE EXCLUSIVE PRICE
    211.00 €
    139.28 € / Pack of 30
    Save 71.72 €
    33% Off
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For use in endotoxin detection assays. Lonza Pyrogen-free Test Tubes without Screw Caps contains less than 0.005 EU/mL endotoxin.

  • Made from USP Type 1 flint borosilicate glass
  • Contain less than 0.005 EU/ml endotoxin
  • Recommended for dilution of endotoxin standards and test samples for all endotoxin detection assays
TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 100 mm
Material Borosilicate Glass
Product Type Test Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.