missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Pyrosperse Dispersing Reagent Kit

Product Code. 10725292
Change view
Click to view available options
Quantity:
300 mL/vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10725292 300 mL/vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10725292 Supplier Lonza™ Supplier No. N188

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Concentration 22%
For Use With (Equipment) Pyrogent QCL-1000
Includes 5mL/vial Pyrosperse dispersing reagent
Quantity 300 mL/vial
Product Type Pyrosperse Dispersing Reagent Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.