missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrotinib dimaleate, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Description
Pyrotinib dimaleate (SHR-1258 dimaleate) is a potent and selective EGFR/HER2 dual inhibitor with IC50s of 13 and 38 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Pyrotinib dimaleate |
| CAS | 1397922-61-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C40H39ClN6O11 |
| Quantity | 10 mg |
| Synonym | SHR-1258 dimaleate |
| Solubility Information | H2O : ≥ 106 mg/mL (130.03 mM) ∣DMSO : 100 mg/mL (122.67 mM; Need ultrasonic) |
| SMILES | O=C(NC1=C(OCC)C=C2N=CC(C#N)=C(NC3=CC=C(OCC4=NC=CC=C4)C(Cl)=C3)C2=C1)/C=C/[C@@H]5N(C)CCC5.O=C(O)/C=C\C(O)=O.O=C(O)/C=C\C(O)=O |
| Molecular Weight (g/mol) | 815.22 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction