missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carbonitrile, 99%

Product Code. 11398236
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11398236

Brand: Thermo Scientific Alfa Aesar L15330.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4513-94-4 | C5H4N2 | 92.10 g/mol

Pyrrole-2-carbonitrile is useful in addition reactions and in heterocycle formation. It acts as a heterocyclic building block in synthetic chemistry. Further, it is used in the preparation of 4-iodopyrrole-2-carbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carbonitrile is useful in addition reactions and in heterocycle formation. It acts as a heterocyclic building block in synthetic chemistry. Further, it is used in the preparation of 4-iodopyrrole-2-carbonitrile.

Solubility
Miscible with benzene, dichloromethane, tertrahydrofuran, dimethylformamide and acetonitrile.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4513-94-4
Molecular Formula C5H4N2
Molecular Weight (g/mol) 92.10
MDL Number MFCD00234061
InChI Key BQMPGKPTOHKYHS-UHFFFAOYSA-N
Synonym pyrrole-2-carbonitrile, 2-cyanopyrrole, unii-6d25bmc1pp, 2-carbonitrilepyrrole, 6d25bmc1pp, cyanopyrrole, 2-cyano-pyrrol, pyrrole carbonitrile, 1h-pyrrolecarbonitrile, 1h-pyrrol-2-kohlenitrile
PubChem CID 138277
IUPAC Name 1H-pyrrole-2-carbonitrile
SMILES N#CC1=CC=CN1

Specifications

Density 1.086
Boiling Point 92°C to 94°C (2 mmHg)
Refractive Index 1.55
Quantity 1 g
UN Number UN3276
Beilstein 107047
Sensitivity Air and light sensitive
Solubility Information Miscible with benzene,dichloromethane,tertrahydrofuran,dimethylformamide and acetonitrile.
Formula Weight 92.1
Percent Purity 99%
Chemical Name or Material Pyrrole-2-carbonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.