missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxaldoxime, 97%, Thermo Scientific™

Product Code. 11399555
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399555 1 g 1g
11309565 5 g 5g
11319565 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399555 Supplier Thermo Scientific Alfa Aesar Supplier No. L18959.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32597-34-5
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00965095
InChI Key WJMIDGRLWRUSSN-UHFFFAOYSA-N
Synonym 1h-pyrrole-2-carbaldehyde oxime, 1h-pyrrole-2-carboxaldehyde oxime, n-1h-pyrrol-2-ylmethylidene hydroxylamine, e-n-1h-pyrrol-2-ylmethylidene hydroxylamine, n-pyrrol-2-ylidenemethyl hydroxylamine, z-n-1h-pyrrol-2-ylmethylidene hydroxylamine
PubChem CID 6743783
IUPAC Name N-(pyrrol-2-ylidenemethyl)hydroxylamine
SMILES C1=CC(=CNO)N=C1

Specifications

Melting Point 163°C to 166°C
Quantity 1 g
Beilstein 108674
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material Pyrrole-2-carboxaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.