missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-3-carboxylic acid, 98+%

Product Code. 15481598
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481598 250 mg 250mg
15461598 1 g 1g
15471598 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481598 Supplier Thermo Scientific Alfa Aesar Supplier No. H55383.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 931-03-3 | C5H5NO2 | 111.1 g/mol

Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.

Solubility
Soluble in water.

Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-03-3
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00800594
InChI Key DOYOPBSXEIZLRE-UHFFFAOYSA-N
Synonym pyrrole-3-carboxylic acid, pyrrole-3-carboxylicacid, 3-carboxy-1h-pyrrole, 1h-pyrrole-3-carboxylicacid, pubchem8345, 3-carboxypyrrole, 3-pyrrolecarboxylic acid, acmc-1agqb, 3-pyrrole-carboxylic acid, pyrrole 3-carboxylic acid
PubChem CID 101030
ChEBI CHEBI:68076
IUPAC Name 1H-pyrrole-3-carboxylic acid
SMILES C1=CNC=C1C(=O)O

Specifications

Quantity 250 mg
Beilstein 108642
Solubility Information Soluble in water.
Formula Weight 111.1
Percent Purity ≥98%
Chemical Name or Material Pyrrole-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.