missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine-1-carbonitrile, 97%, Thermo Scientific™

Product Code. 10686893
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10686893 10 g Amber Glass Bottle 10g
10082882 1 g Amber Glass Bottle 1g
10225944 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10686893 Supplier Thermo Scientific Maybridge Supplier No. SB00582EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1530-88-7
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00003165
InChI Key QJRYYOWARFCJQZ-UHFFFAOYSA-N
Synonym 1-pyrrolidinecarbonitrile, 1-cyanopyrrolidine, n-cyanopyrrolidine, pyrrolidinecarbonitrile, cyanopyrrolidine, cyano-pyrrolidine, tetramethylenecyanamide, 1-pyrrolidine carbonitrile, d00fga
PubChem CID 73737
IUPAC Name pyrrolidine-1-carbonitrile
SMILES N#CN1CCCC1

Specifications

CAS Min % 97.0
Density 0.9540g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 96.13
Percent Purity 97%
Chemical Name or Material Pyrrolidine-1-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.