missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine-1-carboximidamide hydroiodide, 97%, Thermo Scientific™

Product Code. 10653403
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10653403 1 g Amber Glass Bottle 1g
10653593 10 g Amber Glass Bottle 10g
10719615 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10653403 Supplier Thermo Scientific Maybridge Supplier No. KM00683DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 102392-83-6
Molecular Formula C5H12IN3
Molecular Weight (g/mol) 241.076
MDL Number MFCD00114285
InChI Key XPASGQSDLPDWNV-UHFFFAOYSA-N
Synonym pyrrolidine-1-carboximidamide hydroiodide, pyrrolidinecarboxamidine, iodide, c5h11n3.hi
PubChem CID 2794596
IUPAC Name pyrrolidine-1-carboximidamide;hydroiodide
SMILES C1CCN(C1)C(=N)N.I

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 241.07
Percent Purity 97%
Chemical Name or Material Pyrrolidine-1-carboximidamide hydroiodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.