missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, Extra Pure, SLR, Fisher Chemical

Product Code. 10336390 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10336390 100 mL Plastic Coated Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10336390 Supplier Fisher Chemical Supplier No. P/8140/PB05
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 123-75-1
Melting Point -63°C
Color Colorless
pH 12.9
Boiling Point 86°C
Packaging Plastic Coated Glass bottle
Molecular Formula C4H9N
MDL Number MFCD00005249
Quantity 100 mL
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
InChI Key RWRDLPDLKQPQOW-UHFFFAOYSA-N
SMILES C1CCNC1
IUPAC Name pyrrolidine
Molecular Weight (g/mol) 71.12
PubChem CID 31268
ChEBI CHEBI:33135
Formula Weight 71.11g/mol
Vapor Pressure 65mbar at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pyrrolidine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.