missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic acid, 98%

Product Code. 11471558
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471558 2500 g 2500g
11451558 100 g 100g
11461558 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471558 Supplier Thermo Scientific Alfa Aesar Supplier No. A13875.0E
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 127-17-3 | C3H4O3 | 88.06 g/mol

Pyruvic acid is used as a component in culture broths and media as commercial red seaweed polysaccharide. It is involved in the construction of amino acid alanine as well as supplies energy to living cells via citric acid cycle (Krebs cycle). It reacts with N-acetyl mannosamine by an aldol-type condensation to prepare sialic acid. It is employed to study the cultivation of soil bacteria as micro colonies using soil substrate membrane system. It finds application in liquid chromatography and in the determination of organic acids in red wine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvic acid is used as a component in culture broths and media as commercial red seaweed polysaccharide. It is involved in the construction of amino acid alanine as well as supplies energy to living cells via citric acid cycle (Krebs cycle). It reacts with N-acetyl mannosamine by an aldol-type condensation to prepare sialic acid. It is employed to study the cultivation of soil bacteria as micro colonies using soil substrate membrane system. It finds application in liquid chromatography and in the determination of organic acids in red wine.

Solubility
Miscible with chloroform and methanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-17-3
Molecular Formula C3H4O3
Molecular Weight (g/mol) 88.06
MDL Number MFCD00002585
InChI Key LCTONWCANYUPML-UHFFFAOYSA-N
Synonym pyruvic acid, pyroracemic acid, 2-oxopropionic acid, acetylformic acid, alpha-ketopropionic acid, 2-ketopropionic acid, 2-oxopropanoate, propanoic acid, 2-oxo, 2-oxo-propionic acid, 2-oxopropansaeure
PubChem CID 1060
ChEBI CHEBI:32816
IUPAC Name 2-oxopropanoic acid
SMILES CC(=O)C(O)=O

Specifications

Melting Point 11°C to 12°C
Density 1.267
pH 1.2
Boiling Point 164°C to 166°C
Flash Point 83°C (181°F)
Assay Percent Range 98%
Odor Pungent
Refractive Index 1.4295
Quantity 2500 g
UN Number UN3265
Beilstein 506211
Merck Index 14,8021
Solubility Information Miscible with chloroform and methanol.
Formula Weight 88.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Pyruvic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.