missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Acid Ethyl Ester, TRC

Product Code. 30410020
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410020 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410020 Supplier Toronto Research Chemicals Supplier No. TRCP998915500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyruvic Acid Ethyl Ester
CAS 617-35-6
Molecular Formula C5H8O3
Quantity 500 g
Synonym 2-Oxopropanoic Acid Ethyl Ester, 2-Oxopropionic Acid Ethyl Ester, Ethyl 2-Oxopropanoate, Ethyl 2-Oxopropionate, Ethyl Methylglyoxylate, Ethyl Pyruvate, NSC 48386
InChI Formula InChI=1S/C5H8O3/c1-3-8-5(7)4(2)6/h3H2,1-2H3
SMILES CCOC(=O)C(=O)C
IUPAC Name ethyl 2-oxopropanoate
Molecular Weight (g/mol) 116.12
Formula Weight 116.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.