missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic acid, MedChemExpress

Product Code. 16442718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442718 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442718 Supplier Medchem Express Supplier No. HYY0781
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates, proteins, and fats.

Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates, proteins, and fats.

Specifications

Chemical Name or Material Pyruvic acid
CAS 127-17-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C3H4O3
Quantity 5 g
Synonym Acetylformic acid
Solubility Information H2O : 100 mg/mL (1135.59 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (567.79 mM)
SMILES CC(C(O)=O)=O
Molecular Weight (g/mol) 88.06
Formula Weight 88.06
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.