missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Acid Sodium Salt, TRC

Product Code. 30134447
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134447 25 g 25g
30008340 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134447 Supplier Toronto Research Chemicals Supplier No. TRCP99890025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3 H3 O3 . Na
Molecular Weight (g/mol) 110.04
Synonym 2-Oxopropanoic Acid Sodium Salt (1:1), Propanoic acid, 2-oxo-, sodium salt (9CI), Pyruvic acid, sodium salt (7CI,8CI), Sodium 2-oxopropanoate, Sodium Pyruvate, Sodium α-ketopropionate, N-acetyl-D-neuraminic acid (NANA), Sodium Pyruvate, N-acetyl-D-mannosamine (NAM), Pyruvate
IUPAC Name sodium;2-oxopropanoate
SMILES [Na+].CC(=O)C(=O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1
Formula Weight 109.998
Chemical Name or Material Pyruvic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.