missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Q-CLASSIC-90 Two-Door Light Gray Safety Storage Cabinets

Code produit. 15860448 Tous les produits asecos Produits
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Load Bearing Capacity:
4 x 60 kg/19 L Drawers
4 x 60 kg/29 L Drawers
75 kg Shelves, 75 kg Insert, 33 L Sump
missing translation for 'unitSize'
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Load Bearing Capacity unitSize
15860448 75 kg Shelves, 75 kg Insert, 33 L Sump Each
15870448 4 x 60 kg/29 L Drawers Each
15880448 4 x 60 kg/19 L Drawers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 15860448 missing translation for 'mfr' Asecos™ missing translation for 'supplierNo' 3000104133635

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

For the safe and approved storage of flammable hazardous substances in working areas.

  • Extremely robust design with scratch-proof surface
  • Triple-hinged doors
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • Integrated transport base for simple in-house transport
  • Cabinet can be transported upright through doors
  • Adjustable feet in base allow the safe and fast compensation of minor surface unevenness
  • Modern bicolor design - choice of seven different door colors
  • Language-free labelling for optimum international usability
  • Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
  • Locking status indicator (red/green) signals the locking status
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • In accordance with EN 14470-1 and TRGS 510
  • Fire resistance of 90 minutes

Spécification

Certifications/Compliance EN 14470-1
Dimensions (D x W x H) 615 x 1193 x 1953 mm
Door Style Triple-hinged
For Use With (Application) Storage of flammable hazardous substances
Load Bearing Capacity 75 kg Shelves, 75 kg Insert, 33 L Sump
No. of Doors 2
Weight (Metric) 474.5 kg
Color Anthracite Gray, Light Gray
Material Gypsum
Product Type Flammables Storage Cabinet
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.