missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Q-PEGASUS-90 One-Door Yellow Safety Storage Cabinets

Product Code. 15765908 Shop All asecos Products
Change view
Click to view available options
Load Bearing Capacity:
Shelves 75 kg, Insert 75 kg, Sump 22 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Load Bearing Capacity unitSize
15765908 Shelves 75 kg, Insert 75 kg, Sump 22 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15765908 Supplier Asecos™ Supplier No. 3007404033600

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

For the safe and approved storage of flammable hazardous substances in working areas.

  • Extremely robust design with scratch-proof surface
  • Triple-hinged doors
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • One-hand operation for double-door cabinets
  • Container remains in one hand whilst opening
  • Doors remain open in the serial door open arrest system, convenient and safe removal or storage of containers
  • Doors automatically close after approx. 60 seconds
  • Acoustic and visual signal before door closing
  • Modern bicolor design - choice of seven different door colors
  • Language-free labelling for optimum international usability
  • Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
  • Locking status indicator (red/green) signals the locking status
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • In accordance with EN 14470-1 and TRGS 510
  • Fire resistance of 90 minutes
  • Integrated transport base with removable, optional base cover
  • Easy alignment – adjusting aids to compensate for uneven floors

Specifications

Certifications/Compliance EN 14470-1
Configuration 3 x 75 kg shelves; 1 x 75 kg perforated insert; 1 x 22 L bottom collecting sump
Dimensions (D x W x H) 615 x 599 x 1953 mm
Door Style Auto-close
For Use With (Application) Storage of flammable hazardous substances
Load Bearing Capacity Shelves 75 kg, Insert 75 kg, Sump 22 L
No. of Doors 1
Weight (Metric) 314.5 kg
Color Anthracite Gray, Warning Yellow
Material Gypsum
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.