missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ Q-PEGASUS-90 Two-Door Light Gray Safety Storage Cabinets
Shop All asecos Products
Click to view available options
Load Bearing Capacity:
Drawers 60 kg/29 L each
Unit Size:
Each
Description
- Extremely robust design with scratch-proof surface
- Triple-hinged doors
- Safety elements assembled outside the storage compartment for increased protection against corrosion
- One-hand operation for double-door cabinets
- Container remains in one hand whilst opening
- Doors remain open in the serial door open arrest system, convenient and safe removal or storage of containers
- Doors automatically close after approx. 60 seconds
- Acoustic and visual signal before door closing
- Modern bicolor design - choice of seven different door colors
- Language-free labelling for optimum international usability
- Doors lockable with cylinder lock, cabinets can be integrated by the customer in an existing master key system
- Locking status indicator (red/green) signals the locking status
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Visual inspection of the exhaust air function in the exhaust grille (rotating fan impeller)
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- In accordance with EN 14470-1 and TRGS 510
- Fire resistance of 90 minutes
- Integrated transport base with removable, optional base cover
- Easy alignment – adjusting aids to compensate for uneven floors
Specifications
Specifications
| Certifications/Compliance | EN 14470-1 |
| Configuration | 6 x 60 kg/29 L Drawers |
| Dimensions (D x W x H) | 615 x 1193 x 1953 mm |
| Door Style | Auto-close |
| For Use With (Application) | Storage of flammable hazardous substances |
| Load Bearing Capacity | Drawers 60 kg/29 L each |
| No. of Doors | 2 |
| Weight (Metric) | 540 kg |
| Color | Anthracite Gray, Light Gray |
| Material | Gypsum |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction