missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Q-PHOENIX-90 Yellow Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 90 minutes.
Brand: asecos™ 3214904033649
Additional Details : Weight : 485.30521kg
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Space saving folding doors: less required space in front of the cabinet while doors are open
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications
Safety storage cabinet | |
90 mm | |
1050 mm | |
537 kg | |
2 | |
6 x Drawers 60 kg/29 L each | |
1647 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
615 x 1193 x 1953 mm | |
522 mm | |
6 x drawer (sheet steel powder-coated) |
Steel | |
Warning Yellow, Anthracite Gray | |
1193 mm | |
Powder-coated textured | |
Q90.195.120.FD | |
Entry height transport base | |
1953 mm | |
Folding | |
522 x 1050 x 1647 mm | |
615 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |