missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Q-PHOENIX-90 Yellow Safety Storage Cabinets

Product Code. 15987406 Shop All asecos Products
Change view
Click to view available options
Configuration:
3 Shelves, 1 Insert, 1 Bottom Collecting Sump
3 Tray Shelves, 1 Perforated Insert (365 mm), 1 Bottom Collecting Sump
4 Drawers
6 Drawers
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Unit Size
15987406 3 Tray Shelves, 1 Perforated Insert (365 mm), 1 Bottom Collecting Sump Each
15890238 3 Shelves, 1 Insert, 1 Bottom Collecting Sump Each
15937416 4 Drawers Each
15957416 6 Drawers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15987406 Supplier asecos™ Supplier No. 3214904033640
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the safe and approved storage of flammable hazardous substances in working areas with fire resistance up to 90 minutes.

  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Space saving folding doors: less required space in front of the cabinet while doors are open
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 3 Tray Shelves, 1 Perforated Insert (365 mm), 1 Bottom Collecting Sump
Depth (Metric) Exterior 615 mm
Depth (Metric) Interior 522 mm
Dimensions (D x W x H) Interior 522 x 1050 x 1647 mm
Dimensions (D x W x H) 615 x 1193 x 1953 mm
Door Style Folding
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1953 mm
Height (Metric) Interior 1647 mm
Includes Entry height transport base
Model Q90.195.120.FD
No. of Doors 2
No. of Shelves 3
Shelf Type Tray shelf
Surface Coating Powder-coated textured
Weight (Metric) 480.50 kg
Width (Metric) Exterior 1193 mm
Width (Metric) Interior 1050 mm
Color Anthracite Gray, Warning Yellow
Height (Metric) Opening 90 mm
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.