missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Q Sepharose™ High Performance Chromatography Media

Product Code. 11560804 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
75 mL
Unit Size:
75mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11560804 75 mL 75mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11560804 Supplier Cytiva Supplier No. 17101401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Afficher la politique du retour.

Designed for high intermediate and polishing steps in downstream processing

  • Composed of crosslinked agarose beads with mean diameter of 34μm, modified with quaternary (Q) strong anion exchange groups
  • Hydrophilic nature of base matrix ensures low levels of non-specific binding
  • High resolution, high capacity separation with high recovery
  • Chemical stability for effective CIP/sanitization
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

TRUSTED_SUSTAINABILITY

Spécification

Quantity 75 mL
Stability Stable in commonly used aqueous Buffers: 8 M Urea, 6 M Guanidine HCl, 70% Ethanol, 1 M NaOH, 1 M Acetic Acid, 30% Isopropanol, 30% Acetonitrile, 2% SDS
For Use With (Application) For Intermediate PurifiCatIon and Polishing AppliCatIons
Storage Requirements 4°C to 30°C
Product Type Q Sepharose High Performance Media
Flow Rate 30 to 150 cm/Hoμr
Particle Size 24 to 44 μm
Height (English) Bed 3.93 in.
Height (Metric) Bed 10 cm
Ionic Capacity 0.14 to 0.20 mM Cl-/mL
Ion Exchange Type Strong AnIon Exchanger
Max. Pressure <3 bar
Matrix 6% Cross-Linked Agarose
pH Range 1 to 14
Storage Buffer 20% Ethanol
Ligand Type Quaternary Amine
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Q sepharose high performance
Mention d'avertissement
  • Warning
Catégorie de danger
  • Flammable liquid Category 3
Mention de danger
  • H226-Flammable liquid and vapour.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informations supplémentaires
  • MIXTURE LIST-Contain: ethanol
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.