missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Q-VD-OPH, MP Biomedicals™

Product Code. 11465386 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
Unit Size:
1mg
3mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465386 1 mg 1mg
11400895 3 mg 3mg
11410895 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465386 Supplier MP Biomedicals™ Supplier No. 03OPH10901
  • ONLINE EXCLUSIVE PRICE
    386.00 €
    365.40 € / 1mg
    Save 20.60 €
    5% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifically designed for in-vivo research. Q-VD-OPH, MP Biomedicals is synthesized using proprietary technology as a cell-permeable, irreversible caspase inhibitor. The OPH trap, with its superior potency, proven cell permeability and minimal toxicity, p

The Q-VD-OPH irreversibily binds to activated Caspases to block apoptosis. Apoptosis is a physiological form of cell death, which plays an important role in embryogenesis, cellular homoestasis, tissue atrophy, and removal of damaged and mutated cells.

Caspases, which act as molecular chainsaw, are the central components of apoptosis response. Caspases are Cysteine proteases that cleave after aspartate residue in their substrates. At least 7 of 14 known mammalian caspases have important role in apoptosis.

  • Exceptional alternative to the Fluoromethylketone family of inhibitors (these compounds form the basis for important new drug candidates for a wide variety of disease targets)
  • Nontoxic&mdash:preliminary study showed no evidence of toxicity in mice injected intraperitoneally with more than 1000mg/kg
  • Improved potency—IC50 values against purified caspases are in the nanomolar range; in an in vitro cell based assay, greater than 90% inhibition of apoptosis was achieved)
  • Optimal dilutions should be determined by the laboratory for each specific application

Specifications

Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 513.49
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.