missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ QB Series Safety Cover

Product Code. 11440968
Click to view available options
For Use With (Equipment):
4 Block Systems
For 1 Block Systems
For 2 Block Systems
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11440968

Brand: Grant Instruments™ QBL1

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made from tough clear acrylic for maximum visibility. Grant Instruments™ QB Series Safety Covers prevent accidental touching of a hot block or contamination of samples from splashes.

  • For use with QB Series Dry Block Heaters
  • Acrylic construction provides increased visibility
  • Prevents burning from accidentally touching the hot block
  • Clearance height of 85mm
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
For Use With (Equipment) For 1 Block Systems
Material Acrylic
Type Safety Cover
For Use With (Application) For maximum visibility whilst preventing accidental touching of a hot block or contamination of samples from splashes
Height (Metric) 85 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.