missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QSY™ 21 Carboxylic Acid, Succinimidyl Ester

Product Code. 11513667
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11513667 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11513667 Supplier Invitrogen™ Supplier No. Q20132
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

QSY™ 21 carboxylic acid, succinimidyl ester

The amine-reactive quencher, QSY™ 21 succinimidyl ester has a broad and intense absorption (∼661 nm maxima) but no fluorescence making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Wet Ice
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye QSY™ 21
Label Type Quencher Dyes
Product Line QSY

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.