missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Quant-iT™ 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR)

Product Code. 15870210 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kit
1000 assays
Assay:
1 x dsDNA, broad range
1 x dsDNA, high sensitivity
Quantitation Range:
0.2 to 100 ng
4 to 4000 ng
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Assay Quantitation Range Unit Size
15870210 1000 assays 1 x dsDNA, high sensitivity 0.2 to 100 ng Each
17116039 1 kit 1 x dsDNA, broad range 4 to 4000 ng Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15870210 Supplier Invitrogen™ Supplier No. Q33232
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For easy and accurate dsDNA quantitation, the Quant-iT 1X dsDNA HS and BR Assays enable up to 1,000 fluorescence-based assays in a 96-well microplate format.

Make DNA quantitation easy and accurate in a ready-to-go 1X formulation with the Quant-iT 1X dsDNA HS (High-Sensitivity) and Quant-iT 1X dsDNA BR (Broad Range) Assay kits. Both kits provide a ready-to-use working solution. Simply load your sample, add the Qubit 1X dsDNA HS or BR working solution, and read the concentration using a fluorescence-based microplate reader.

The Quant-iT 1X dsDNA HS and BR Assay kits are highly selective for double-stranded DNA (dsDNA) over RNA, providing an accurate and precise method for the quantification of dsDNA samples. To perform the assay, dilute your sample (any volume from 1–20 μL is acceptable) into the 1X working solution provided, then read the concentration using a fluorescence plate reader.

The Quant-iT 1X dsDNA HS Assay Kit is accurate for initial sample concentrations ranging from 10 pg/μL to 100 ng/μL, providing a core detection range of 0.2 ng to 100 ng of DNA in the assay tube. The Quant-iT 1X dsDNA BR Assay Kit is accurate for initial DNA sample concentrations of 200 pg/μL to 4000 ng/μL (depending on sample volume), providing a detection range of 4–4000 ng of DNA.

Common contaminants such as salts, free nucleotides, solvents, detergents, and protein are well tolerated in the dsDNA assays. The '1X' dsDNA assay kits provide reagent and buffer in a ready-to-use formulation and are highly selective for dsDNA over ssDNA, RNA, and protein.
TRUSTED_SUSTAINABILITY

Specifications

Assay 1 x dsDNA, high sensitivity
Detection Method Fluorescence
Format Kit
Sample Type DNA
For Use With (Equipment) Microplate Reader
Quantitation Range 0.2 to 100 ng
Product Line Quant-iT
Quantity 1000 assays
Shipping Condition Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.