missing translation for 'onlineSavingsMsg'
Learn More

QuantiGene™ Plex Wash Buffer Components

Product Code. 32045483
Change view
Click to view available options
Quantity:
1 Plate
10 Plates
3 Plates
Unit Size:
Each
Pack of 10
Pack of 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32045483 3 Plates Pack of 3
32044532 1 Plate Each
32044931 10 Plates Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32045483 Supplier Invitrogen Supplier No. QP1502
  • ONLINE EXCLUSIVE PRICE
    103.00 €
    83.43 € / Pack of 3
    Save 19.57 €
    19% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer.

QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer. The wash buffer components may be purchased in 1, 3, or 10-plate quantities to supplement QuantiGene Plex Assay kits when additional wash buffer volume is required for automation.

TRUSTED_SUSTAINABILITY

Specifications

Format 96-well
For Use With (Application) QuantiGene Plex Assay
Content And Storage • 2.4 mL Wash Buffer Component 1
• 39 mL Wash Buffer Component 2

Store at room temperature.

Key Functions Multi-Gene Expression
Product Line QuantiGene Plex
Product Type Wash Buffer
Quantity 3 Plates
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.